Documents
WHA10328170
Whatman® cotton grade 740E filter discs
white, hydrophilic, thick, 1000 ea, 12.7 mm diam
Synonyme(s) :
Z749486
About This Item
Code UNSPSC :
40161507
Nomenclature NACRES :
NB.24
product name
Whatman® filter discs, Grade 740E, size 12.7 mm
Stérilité
non-sterile
Conditionnement
pk of 1000
Fabricant/nom de marque
Whatman 10328170
Whatman Article No. 28414439 (US reference)
Taille
12.7 mm
Description générale
Whatman® Grade 740E Filter Discs are thick, and highly absorbent paper discs made of high-purity cotton linter and are typically used in antibiotic-assay procedures.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva