Principaux documents
SYNF212500
Synthware four-neck round-bottom flask with vertical side necks
capacity 500 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Synonyme(s) :
Synthware F212500, 4-neck reaction flask
1 EA
142,00 €
142,00 €
Check Cart for Availability
Changer de vue
1 EA
142,00 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
142,00 €
Check Cart for Availability
Matériaux
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS29/42
side joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F212500
Capacité
500 mL
Description générale
With identical vertical side joints.
Informations légales
Synthware is a trademark of Kemtech America Inc.