Principaux documents
DWK11-5300-500
Wheaton® Microliter Vial
1 PKG
55,10 €
55,10 €
Check Cart for Availability
Changer de vue
1 PKG
55,10 €
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.11
55,10 €
Check Cart for Availability
Matériaux
conical polypropylene vial
Conditionnement
pkg of 1 packs
Fabricant/nom de marque
WHEATON® 11-5300-500
Capacité
500 μL
Application
Chromatography
Caractéristiques et avantages
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.