Principaux documents
CLS566050
Pyrex® Lifetime-Red volumetric flask with Pyrex® stopper
capacity 50 mL
Synonyme(s) :
flasks glass, volumetric flasks, volumetric glass, Corning® 5660
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass
long
Joint
joint stopper9
Stérilité
non-sterile
Caractéristiques
closure type ST stopper
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 5660-50
tol. measuring range
0.05 mL
Capacité
50 mL
Caractéristiques et avantages
  • Pyrex® Lifetime-Red volumetric flasks with Pyrex® stoppers (volume 50 mL) have the capacity mark etched as a fine line through a narrow red band into clear glass, making a filler unnecessary and giving high legibility, accuracy and permanence.
  • The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for No. 9 Standard Taper glass stoppers.
  • These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware and ASTM E-288 for volumetric flasks. Calibrated.
Informations légales
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.