Skip to Content
Merck
All Photos(1)

Documents

58722

Tubing Adapter

1/4-28 male UNF, male Luer

Synonym(s):

Male Luer Adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815

material

CTFE

description

for chromatography applications

packaging

pkg of 1 ea

manufacturer/tradename

VICI Valco® CMLAKF

fitting

male Luer
1/4-28 male UNF

Looking for similar products? Visit Product Comparison Guide

General description

Male adapter connects needle or Luer hub to ¼"-28 threaded tubing or adapter. Order coupling separately.
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It may be used to connect capillary tubes in HPLC, GC etc.

Application

Tubing adaptor was used in solid phase extraction.

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

C Brede et al.
Food additives and contaminants, 21(11), 1115-1124 (2005-03-15)
Migration of aniline from polyamide cooking utensils was investigated. Aniline was found to originate from the application of black colorant in the polyamide 66 raw material. Analysis of polyamide raw material revealed an aniline concentration of 121 +/- 13 mg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service