Documents
55071
Tubing Adapter
1/4-28 female (flat-bottom), 10-32 male (standard)
About This Item
UNSPSC Code:
41115700
material
PEEK adapter (one-piece design)
description
for chromatography applications
packaging
pkg of 1 ea
bore diam.
0.030 in.
fitting
1/4-28 female (flat-bottom)
10-32 male (standard)
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)