Skip to Content
Merck
All Photos(1)

Key Documents

30-3815

Sigma-Aldrich

Tributyl phosphate

SAJ first grade, ≥99.0%

Synonym(s):

TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

vapor density

9.2 (vs air)

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

770 °F

availability

available only in Japan

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

294.8 °F

Flash Point(C)

146 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C R Deeken et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 96(2), 199-206 (2011-01-07)
Extracellular matrix (ECM) materials are currently utilized for soft tissue repair applications such as vascular grafts, tendon reconstruction, and hernia repair. These materials are derived from tissues such as human dermis and porcine small intestine submucosa, which must be rendered
Muniasamy Neerathilingam et al.
Toxicology letters, 199(1), 10-16 (2010-08-07)
Tributyl phosphate (TBP) is a toxic organophosphorous compound widely used in many industrial applications, including significant usage in nuclear processing. The industrial application of this chemical is responsible for occupational exposure and environmental pollution. In this study, (1)H NMR-based metabonomics
C W Van Neste et al.
Analytical chemistry, 81(5), 1952-1956 (2009-02-04)
Despite its immediate applications, selective detection of trace quantities of surface adsorbed chemicals, such as explosives, without physically collecting the sample molecules is a challenging task. Standoff spectroscopic techniques offer an ideal method of detecting chemicals without using a sample
Domenico Santoro et al.
Environmental science & technology, 44(16), 6233-6241 (2010-08-14)
Advanced Oxidation Processes (AOPs) promoted by ultraviolet light are innovative and potentially cost-effective solutions for treating persistent pollutants recalcitrant to conventional water and wastewater treatment. While several studies have been performed during the past decade to improve the fundamental understanding
Torsten Klengel et al.
Nature neuroscience, 16(1), 33-41 (2012-12-04)
Although the fact that genetic predisposition and environmental exposures interact to shape development and function of the human brain and, ultimately, the risk of psychiatric disorders has drawn wide interest, the corresponding molecular mechanisms have not yet been elucidated. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service