Merck
  • Allele-specific FKBP5 DNA demethylation mediates gene-childhood trauma interactions.
Allele-specific FKBP5 DNA demethylation mediates gene-childhood trauma interactions.
Nature neuroscience (2012-12-04)
Torsten Klengel, Divya Mehta, Christoph Anacker, Monika Rex-Haffner, Jens C Pruessner, Carmine M Pariante, Thaddeus W W Pace, Kristina B Mercer, Helen S Mayberg, Bekh Bradley, Charles B Nemeroff, Florian Holsboer, Christine M Heim, Kerry J Ressler, Theo Rein, Elisabeth B Binder
ABSTRACT
Although the fact that genetic predisposition and environmental exposures interact to shape development and function of the human brain and, ultimately, the risk of psychiatric disorders has drawn wide interest, the corresponding molecular mechanisms have not yet been elucidated. We found that a functional polymorphism altering chromatin interaction between the transcription start site and long-range enhancers in the FK506 binding protein 5 (FKBP5) gene, an important regulator of the stress hormone system, increased the risk of developing stress-related psychiatric disorders in adulthood by allele-specific, childhood trauma-dependent DNA demethylation in functional glucocorticoid response elements of FKBP5. This demethylation was linked to increased stress-dependent gene transcription followed by a long-term dysregulation of the stress hormone system and a global effect on the function of immune cells and brain areas associated with stress regulation. This identification of molecular mechanisms of genotype-directed long-term environmental reactivity will be useful for designing more effective treatment strategies for stress-related disorders.
Product Description
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tributyl phosphate, ≥99%
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
Sigma-Aldrich
Tributyl phosphate, 97%