Skip to Content
Merck
All Photos(1)

Documents

07-0635

Sigma-Aldrich

Devarda′s alloy

for determination of nitrogen

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlCuZn
CAS Number:
MDL number:
UNSPSC Code:
12352402
PubChem Substance ID:

grade

for determination of nitrogen

form

powder

composition

aluminum, ~45%
copper, ~50%
zinc, ~5%

availability

available only in Japan

mp

550 °C (lit.)

SMILES string

[Al].[Cu].[Zn]

InChI

1S/Al.Cu.Zn

InChI key

MUBKMWFYVHYZAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service