Skip to Content
Merck
All Photos(1)

Documents

662895

Sigma-Aldrich

2-Hexynoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH≡CCO2H
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.460

bp

230 °C

density

0.992 g/mL at 25 °C

SMILES string

CCCC#CC(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-3H2,1H3,(H,7,8)

InChI key

AKYAUBWOTZJUBI-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louis DeRidder et al.
Nanoscale, 13(2), 939-952 (2021-01-23)
Switching microglia from a disease exacerbating, 'pro-inflammatory' state into a neuroprotective, 'anti-inflammatory' phenotype is a promising strategy for addressing multiple neurodegenerative diseases. Pro-inflammatory microglia contribute to disease progression by releasing neurotoxic substances and accelerating pathogenic protein accumulation. PPARα and PPARγ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service