Skip to Content
Merck
All Photos(1)

Key Documents

PHL84146

Rebaudioside M

phyproof® Reference Substance

Synonym(s):

Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl ester, (4α)-, Rebaudioside X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H90O33
CAS Number:
Molecular Weight:
1291.29
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

Looking for similar products? Visit Product Comparison Guide

General description

Componentof stevia.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Efficient Conversion of Stevioside to Rebaudioside M in Saccharomyces cerevisiae by a Engineering Hydrolase System and Prolonging the Growth Cycle.: This study demonstrates an innovative method for converting Stevioside to high-purity Rebaudioside M using an engineered hydrolase system in yeast, which could significantly impact the production of natural sweeteners for pharmaceutical applications (Li et al., 2024).
  • Acute and two-week effects of neotame, stevia rebaudioside M and sucrose-sweetened biscuits on postprandial appetite and endocrine response in adults with overweight/obesity-a randomised crossover trial from the SWEET consortium.: This randomized crossover trial investigates the short-term and two-week effects of Rebaudioside M on appetite and endocrine response, providing insights into its benefits and applications as a non-caloric sweetener in the pharmaceutical industry (Gibbons et al., 2024).
  • Comparative transcriptomic of Stevia rebaudiana provides insight into rebaudioside D and rebaudioside M biosynthesis.: The research offers a comparative transcriptomic analysis of Stevia rebaudiana, enhancing our understanding of the biosynthesis of Rebaudioside M and its potential applications in glycoside-based drug formulations (Wang et al., 2021).
  • Structural Insights into the Catalytic Mechanism of a Plant Diterpene Glycosyltransferase SrUGT76G1.: This study provides structural insights into the catalytic mechanisms of a key enzyme involved in the production of Rebaudioside M, which is crucial for its biochemistry and applications in high-purity natural sweeteners (Liu et al., 2020).
  • Structural characterization of the degradation products of a minor natural sweet diterpene glycoside Rebaudioside M under acidic conditions.: This article explores the stability and degradation of Rebaudioside M under acidic conditions, important for its formulation and stability in pharmaceutical products (Prakash et al., 2014).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service