Skip to Content
Merck
Rebaudioside M ≥95.0% (HPLC)

Rebaudioside M

Rebaudioside M ≥95.0% (HPLC)
Synonym(s):
Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl ester, (4α)-, Rebaudioside X
Empirical Formula (Hill Notation):
C56H90O33
CAS Number:
Molecular Weight:
1291.29

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing