Skip to Content
Merck
All Photos(1)

Key Documents

171596

Sigma-Aldrich

β-Amyloid Peptide

≥80% (HPLC), lyophilized, 1-42, rat

Synonym(s):

β-Amyloid Peptide (1-42), Rat, DAEFGHDSGFEVRHQKLVFFAEDVGSNKGAIIGLMVGGVVIA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C199H307N53O59S
CAS Number:
Molecular Weight:
4417.95
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

β-Amyloid Peptide (1-42), Rat,

description

Predominant peptide found in the brain of patients with Alzheimer′s Disease and Down′s Syndrome. Promotes down-regulation of Bcl-2 and upregulation of Bax expression in neurons.

Quality Level

Assay

≥80% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(

InChI

1S/C203H311N55O60S/c1-28-106(20)164(195(310)220-91-149(267)228-130(71-98(4)5)181(296)238-129(66-70-319-27)179(294)251-158(100(8)9)193(308)218-87-146(264)215-88-151(269)250-160(102(12)13)198(313)255-163(105(18)19)199(314)258-165(107(21)29-2)200(315)227-112

InChI key

DZHSAHHDTRWUTF-SIQRNXPUSA-N

General description

β-Amyloid Peptide (Aβ) is the main constituent in both senile plaques and diffuse deposits in Alzheimer′s diseased brains. Under appropriate conditions, the soluble amyloid peptides aggregate into the fibrils associated with the disease state. It has proposed that Aβ peptide may increase the sensitivity of neurons to oxidative damage by downregulating expression of bcl-2, a key anti-apoptotic protein and upregulating of bax expression, a protein known to induce cell death.
Predominant peptide found in the brain of patients with Alzheimer’s Disease and Down’s Syndrome. Promotes down-regulation of Bcl-2 and upregulation of Bax expression in neurons.

Warning

Toxicity: Standard Handling (A)

Sequence

H-Asp-Ala-Glu-Phe-Gly-His-Asp-Ser-Gly-Phe-Glu-Val-Arg-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH

Other Notes

Paradis, E., et al. 1996. J. Neurosci.16, 7533.
Roher, A.E., et al. 1996. J. Biol. Chem.271, 20631.
Soto, C., and Castano, E.M. 1996. Biochem. J.314, 701.
Murrell, J., et al. 1991. Science254, 97.
Goldgaber, D., et al. 1987. Science235, 877.
Kang, J., et al. 1987. Nature 325, 733.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service