Skip to Content
Merck
All Photos(1)

Key Documents

L-2

90112811, rat lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

L-2, 90112811

biological source

rat lung

growth mode

Adherent

karyotype

Modal no. 77

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat lung

Cell Line Description

Derived from normal rat lung tissue. The cells possess elevated levels of specific enzymes involved in the synthesis of phosphatidyl choline. Osmiophilic lamellar inclusions are observed in cell pellets from mass culture.

Culture Medium

Coon′s modified Ham′s F12 or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service