Skip to Content
Merck
All Photos(1)

Documents

TA3 Hauschka

85061102, mouse breast, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

TA3 Hauschka, 85061102

biological source

mouse breast

growth mode

Adherent

karyotype

2n = 41

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse mammary carcinoma

Cell Line Description

In form of tumorigenic ascites shows diminished expression of histocompatibility antigens and readily overcomes histoincompatibility barriers. Immunoresistant TA3/Ha sub-line arose spontaneously without subjection to any known immunological selection pressure.

Application

Tumourigenicity studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Cells grow partly in suspension, therefore spin down supernatant at time of subculture to collect the cells.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service