Key Documents
Z688045
CELLine adhere bioreactor flask
AD 1000 (For adherent cell types)
Synonym(s):
Adherent Cell Culture Flask
About This Item
UNSPSC Code:
41122104
NACRES:
NB.22
material
polystyrene (with 100,000 D MWCO regenerated cellulose semi-permeable membrane)
polystyrene
sterility
sterile; γ-irradiated
packaging
case of 3
manufacturer/tradename
Cole Parmer 2440655
capacity
1000 mL
General description
Ideal for MAb and cellular protein production. Viable in vitro alternative to using mouse ascites. Bioreactors will utilize innovative membrane technology to separate the cell cultivation area from the media chamber. Cell and cellular-secreted product with a MWCO of 10 kDa are retained between two non-cytotoxic membranes. The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination. The lower membrane provides direct oxygenation and ensures optimum gas exchange.