Z688037
CELLine Classic bioreactor flask
CL 350 (for 350ml volumes)
Synonym(s):
Bioreactor Vessel, CELLine Flask
About This Item
Recommended Products
material
polystyrene (with 100,000 D MWCO regenerated cellulose semi-permeable membrane)
description
Equivelent to 16 T-225 standard flasks
sterility
sterile; γ-irradiated
usage
Production of monoclonal antibodies and cellular proteins
packaging
case of 5
manufacturer/tradename
Cole Parmer 2440654
capacity
350 mL