PCG2011
TruPAGE™ Precast Gels
4-12%, 10 x 8cm, 12-well
Synonym(s):
Precast Gels for SDS-PAGE, Precast Polyacrylamide Gels
About This Item
Recommended Products
description
for use with Bio-Rad Mini-PROTEAN™ Tetra Cell, Mini-PROTEAN Dodeca™, Mini-PROTEAN II and 3 cells
shelf life
24 mo. at 2‑8 °C (from the date of manufacture)
packaging
box of 10 ea
concentration
4-12% polyacrylamide
cassette W × L
10 cm × 8 cm
gel thickness
1 mm
well size × volume
12 wells × 35 μL
compatibility
for use with Hoefer Mighty Small II and Tall Mighty Small (SE 260 and SE280)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
The gels come in two gel cassette sizes (10 x 10 cm and 10 x 8 cm) for increased equipment compatibility. TruPAGE gels have extra tall wells to prevent lane-to-lane overflow and come in two well formats (12-well and 17-well). TruPAGE gels are available in several different gel concentrations (10%, 12%, 4-8%, 4-12% and 4-20%) to provide desired resolution of proteins of any size. Please refer to the migration chart to select the appropriate precast gel and running buffer combination for your experimental needs. Only use TruPAGE formulated buffers when running TruPAGE gels to ensure optimal results and reproducibility. Please review the TruPAGE Technical Bulletin for buffer formulation recipes and general running guidelines.
- Long Shelf Life - 2 years from the date of manufacture when stored properly.
- High Performance - Extended full length resoling gels provides improved resolution.
- Durable - Stronger than standard polyacrylamide gels, preventing tearing during post-electrophoresis processing.
- Extra Tall Wells - Eliminate sample spill over and accommodate larger sample loading volumes with well teeth that protrude above the back plate of the gel cassette.
- Excellent Compatibility - TruPAGE gels are compatible with the popular gel running equipment, including Sigma-Aldrich′s Dual Run Tank (Z741768) and other manufacturers like Life Technologies, Bio-Rad®, Lonza®, Hoefer®, CBS Scientific as well as others, allowing you to continue using the equipment you already own.
- Fast - Run times as low as 30 minutes.
- Great Value - Gels and buffers offer a cost-saving alternative without sacrificing performance and experimental integrity.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service