Skip to Content
FK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
TruPAGE
™
Precast Gels
Synonym(s):
Precast Gels for SDS-PAGE, Precast Polyacrylamide Gels
Products
Product Category
precast gels (8)
Brand
Sigma-Aldrich (8)
Length (cm)
Volume (µL)
Wells
Compatibility
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
PCG2012
TruPAGE
™
Precast Gels,
4-20%, 10 x 8cm, 12-well
Expand
View Pricing
PCG2003
TruPAGE
™
Precast Gels,
4-12%, 10 x 10cm, 12-well
Expand
View Pricing
PCG2004
TruPAGE
™
Precast Gels,
4-20%, 10 x 10cm, 12-well
Expand
View Pricing
PCG2010
TruPAGE
™
Precast Gels,
12%, 10 x 8cm, 12-well
Expand
View Pricing
PCG2015
TruPAGE
™
Precast Gels,
4-12%, 10 x 8cm, 17-well
Expand
View Pricing
PCG2019
TruPAGE
™
Precast Gels,
4-8%, 10 x 8cm, 12-well
Expand
View Pricing
PCG2011
TruPAGE
™
Precast Gels,
4-12%, 10 x 8cm, 12-well
Expand
View Pricing
PCG2006
TruPAGE
™
Precast Gels,
12%, 10 x 10cm, 17-well
Expand
View Pricing