Skip to Content
Merck
All Photos(1)

Key Documents

SSE-5

95122021, fish embryo, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

SSE-5, 95122021

biological source

fish embryo

packaging

tube of 5 μg 95122021-DNA-5UG
pkg of vial of cells 95122021-1VL

growth mode

Adherent

karyotype

Modal no. 55

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish Sockeye Salmon embryo

Cell Line Description

SSE-5 was initiated in 1964 from pooled embryos of Sockeye Salmon (Oncorynchus nerka). The cell line that is also known as Sockeye Salmon embryo-5, is susceptible to infectious pancreatic necrosis virus (IPNV), viral hemorrhagic septicaemia virus (VHSV) and infectious hematopoietic necrosis virus (IHNV). For replication of salmonid viruses SSE-5 cells should be grown at 18°C.

Application

Isolation of viruses from clinical samples, virus propagation

Culture Medium

MEM (EBSS) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 15-21°C; split every 10-14 days. Fish cell lines detach easily during transit if the culture is very young. After resuscitation split cells 1:3 and culture for at leas

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service