Skip to Content
Merck
All Photos(1)

Key Documents

CSE-119

95122019, fish unknown/unspecified, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CSE-119, 95122019

biological source

fish unknown/unspecified

packaging

tube of 5 μg 95122019-DNA-5UG
pkg of vial of cells 95122019-1VL

growth mode

Adherent

karyotype

Diploid and modal no. 60

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish Coho salmon fibroblast

Cell Line Description

The fish cell line CSE-119 was initiated in 1963 from pooled embryos of Coho salmon (Oncorynchus kisutch). CSE-119, also known as Coho salmon embryo-119, are susceptible to infectious pancreatic necrosis virus (IPNV) but show a lack of sensitivity to infection with viral haemorrhagic septicaemia virus (VHSV) or infectious haematopoietic necrosis virus (IHNV). This might be correlated to the natural resistance to infectious haemotopoietic necrosis in this species. For replication of salmonid viruses CSE-119 cells should be incubated at 18°C.

Application

Isolation of viruses from clinical samples, virus propagation

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 15-21°C; split every 10-14 days; requires a new flask every passage. Fish cell lines detach easily during transit if the culture is too young. After resuscitation spli

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Dr J L Fryer, Department of Microbiology, Oregon State University,Corvallis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service