Skip to Content
Merck
All Photos(1)

Documents

T2813

Millipore

Tryptose

pkg of 500 g, suitable for microbiology

Synonym(s):

Peptone from meat and soybean meal, Tryptic digested peptone

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

bovine milk

Quality Level

form

powder

packaging

pkg of 500 g

storage condition

(Keep dry and tightly closed)

color

light yellow to yellow

pH

6.2-7.2

application(s)

food and beverages
microbiology

General description

Tryptose is a mixture of tryptic-digested proteins. It is a complex mixture of peptides and amino acids obtained from both animal and plant sources, primarily from meat and soybean meal. The formulation of tryptose provides an excellent growth medium, rich in essential nutrients that foster the growth and metabolism of bacteria, yeast, and other fastidious microorganisms.

Application

Tryptose is widely used in the preparation of microbial culture media to grow fastidious organisms. It is extensively used in microbiological research and industrial applications, notably for the maintenance and propagation of various microbial cultures.

Storage and Stability

Keep the container tightly closed in a dry and well-ventilated place

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service