11835289910
Roche
PCR DIG Labeling MixPLUS
sufficient for 2 x 50 assays (50μl final reaction volume), pkg of 2 × 250 μL, solution
Synonym(s):
DIG system, PCR labeling
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105500
Recommended Products
form
solution
usage
sufficient for 2 x 50 assays (50μl final reaction volume)
packaging
pkg of 2 × 250 μL
manufacturer/tradename
Roche
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
, Aligned
storage temp.
−20°C
General description
The PCR DIG Labeling MixPLUS is a mixture of lithium salts of deoxyadenosine triphosphate (dATP), deoxycytosine triphosphate (dCTP), deoxyguanosine triphosphate (dGTP), deoxyuridine triphosphate (dUTP) and digoxigenin (DIG)-11-dUTP. This nucleotide mixture can be added directly to polymerase chain reactions (PCR) and the DIG-labeled nucleotide will be incorporated into the PCR product. The incorporation of dUTP instead of deoxythymidine triphosphate (dTTP) allows the degradation of contaminating PCR products from previous amplification reactions with uracil-DNA glycosylase (UNG), preventing carryover contamination.The incorporated DIG label renders the sensitive detection of the PCR product with anti-DIG-conjugates, for example in the PCR ELISA, DIG-labling, and PCR ELISA, DIG-Detection. The PCR DIG Labeling MixPLUS gives comparable signal intensities to the PCR DIG Labeling Mix.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical. The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.
Application
PCR DIG Labeling MixPLUS is used for direct labeling of amplification products with digoxigenin (DIG)-deoxyuridine triphosphate (dUTP) in polymerase chain reactions (PCR) and for carryover prevention.The PCR DIG Labeling Mixplus is 10x concentrated. For each reaction, add 10% of the final volume to the reaction mix.
Quality
The PCR DIG Labeling MixPLUS is function tested.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service