Skip to Content
Merck
All Photos(1)

Key Documents

AB9206

Sigma-Aldrich

Anti-DARPP-32 Antibody, phosphoThr34

Chemicon®, from rabbit

Synonym(s):

Anti-DARPP-32, Anti-DARPP32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human, mouse, rat
technique(s):
western blot: suitable
citations:
2

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr34)

Gene Information

human ... PPP1R1B(84152)

Specificity

Recognizes DARPP-32 phosphorylated at threonine 34 .

Immunogen

Epitope: phosphoThr34
Synthetic peptide from DARPP-32, phosphoThr34.

Application

Research Category
Neuroscience
Research Sub Category
Reward & Addiction

Signaling Neuroscience
This Anti-DARPP-32 Antibody, phosphoThr34 is validated for use in WB for the detection of DARPP-32.
Western blot: 1:1,000. The antibody recognizes the ~32 kD DARPP-32 protein phosphorylated at Thr34 protein in rat caudate lysate prepared from caudate slices that had been incubated in the absence and presence of forskolin. The labeling by the antibody to DARPP-32 Thr34 is markedly increased by the forskolin treatment.

Optimal working dilutions must be determined by the end user.

Target description

~32 kD

Physical form

Affinity Purified immunoglobulin. Liquid in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg per mL BSA with 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Cultured primary neurons, rat brain lysate and human breast adenocarcinoma

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lídia Cantacorps et al.
British journal of pharmacology, 177(5), 1090-1105 (2019-11-11)
Alcohol exposure in utero may lead to a wide range of long-lasting morphological and behavioural deficiencies known as fetal alcohol spectrum disorders (FASD), associated with a higher risk of later developing neuropsychiatric disorders. However, little is known about the long-term
Maria De Risi et al.
Nature communications, 12(1), 3495-3495 (2021-06-11)
Lysosomal storage disorders characterized by altered metabolism of heparan sulfate, including Mucopolysaccharidosis (MPS) III and MPS-II, exhibit lysosomal dysfunctions leading to neurodegeneration and dementia in children. In lysosomal storage disorders, dementia is preceded by severe and therapy-resistant autistic-like symptoms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service