Skip to Content
Merck
All Photos(2)

Key Documents

364-M

Sigma-Aldrich

Toluidine Blue O

Synonym(s):

Toluidine Blue O, Methylene Blue T

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
305.82
Colour Index Number:
52040
MDL number:
UNSPSC Code:
41116130
EC Index Number:
202-146-2

form

solid

Quality Level

pH

2.8 (25 °C in H2O)

solubility

30 g/L

λmax

620-638 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

SMILES string

[s+]1c2c(nc3c1cc(cc3)N(C)C)cc(c(c2)N)C.[Cl-]

InChI

1S/C15H16N3S.ClH/c1-9-6-13-15(8-11(9)16)19-14-7-10(18(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H/q+1;/p-1

InChI key

HNONEKILPDHFOL-UHFFFAOYSA-M

General description

Toluidine Blue O
364-12 25g Glass Bottle

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
For use in histological sections, animal tisuue and as a general nuclear stain.
HARLECO® For use in histological sections, animal tissue and as a general nuclear stain

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service