Skip to Content
Merck
All Photos(1)

Documents

W254820

Sigma-Aldrich

Heptyl alcohol

natural, 98%

Synonym(s):

1-Heptanol, Heptyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
FEMA Number:
2548
Beilstein:
1731686
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.021
NACRES:
NA.21

grade

Halal
Kosher
natural

reg. compliance

FDA 21 CFR 117

vapor pressure

0.5 mmHg ( 20 °C)

Assay

98%

autoignition temp.

662 °F

refractive index

n20/D 1.424 (lit.)

bp

176 °C (lit.)

mp

−36 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; leafy; waxy; vegetable; fruity

SMILES string

CCCCCCCO

InChI

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural occurrence: Apple, apricot, beer, butter, macadamia nut, mushroom, strawberry, tea, tobacco, and wine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul G Green et al.
The journal of pain : official journal of the American Pain Society, 16(9), 881-886 (2015-06-19)
Corneal injury can produce photophobia, an aversive sensitivity to light. Using topical application of lidocaine, a local anesthetic, and tetrodotoxin (TTX), a selective voltage-sensitive sodium channel blocker, we assessed whether enhanced aversiveness to light induced by corneal injury in rats
Shawn M Stevens et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 150(4), 659-665 (2014-01-18)
Identify cells supporting cochlear lateral wall regeneration. Prospective controlled trial. Laboratory. Human presbyacusis occurs, in part, secondary to age-related degeneration of cochlear lateral wall structures such as the stria vascularis and spiral ligament fibrocytes. This degeneration is likely linked to
Olga A Rozentsvet et al.
Phytochemistry, 105, 37-42 (2014-06-04)
The composition of membrane-forming lipids has been examined for 10 wild halophyte species growing in southern Russian on alkaline soil. The plants belong to seven taxa of family rank: by their life form, which are semi-shrubs, herbaceous annuals, and perennial
Isobel Massie et al.
Experimental eye research, 127, 196-205 (2014-08-12)
Limbal epithelial stem cell deficiency can cause blindness but may be treated by human limbal epithelial cell (hLE) transplantation, normally on human amniotic membrane. Clinical outcomes using amnion can be unreliable and so we have developed an alternative tissue equivalent
Saeed Nojavan et al.
Analytica chimica acta, 838, 51-57 (2014-07-30)
There are numerous published reports about dispersive liquid phase microextraction of the wide range of substances, however, till now no broadly accepted systematic and purpose oriented selection of extraction solvent has been proposed. Most works deal with the optimization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service