Skip to Content
Merck
Heptyl alcohol natural, 98%

Heptyl alcohol

Heptyl alcohol natural, 98%
Synonym(s):
1-Heptanol, Heptyl alcohol
Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing