Skip to Content
Merck
All Photos(1)

Key Documents

360015

Sigma-Aldrich

tert-Butyl methyl malonate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2COOC(CH3)3
CAS Number:
Molecular Weight:
174.19
Beilstein:
1772136
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

impurities

5% dimethyl malonate

refractive index

n20/D 1.415 (lit.)

bp

80 °C/11 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)CC(=O)OC(C)(C)C

InChI

1S/C8H14O4/c1-8(2,3)12-7(10)5-6(9)11-4/h5H2,1-4H3

InChI key

XPSYZCWYRWHVCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl malonate is an ester. Stereospecific Pd(O)-catalyzed addition of tert-butyl methyl malonate to allylic carbonates is reported.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingli Yan et al.
The Journal of organic chemistry, 69(8), 2859-2862 (2004-04-13)
A combination of cross-metathesis and malonate addition was applied to a formal synthesis of the mammalian lignan enterolactone 5. The cross-metathesis of alkene 6 and phosphonate 3a gave the substituted allylic phosphonate 3d. The palladium-catalyzed addition of malonate 10d to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service