Skip to Content
Merck
tert-Butyl methyl malonate 95%

tert-Butyl methyl malonate

tert-Butyl methyl malonate 95%
Linear Formula:
CH3OCOCH2COOC(CH3)3
CAS Number:
Molecular Weight:
174.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing