About This Item
Recommended Products
isotopic purity
98 atom % D
Assay
99% (CP)
form
solid
mp
117-119 °C (lit.)
mass shift
M+4
SMILES string
[2H]c1c([2H])c(C=O)c([2H])c([2H])c1O
InChI
1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H/i1D,2D,3D,4D
InChI key
RGHHSNMVTDWUBI-RHQRLBAQSA-N
Related Categories
1 of 4
This Item | 614726 | 485233 | 588652 |
---|---|---|---|
98 atom % D | 98 atom % D | 99 atom % 13C | 99 atom % D |
M+4 | M+5 | M+1 | M+6 |
solid | solid | solid | solid |
117-119 °C (lit.) | 117-119 °C (lit.) | 117-119 °C (lit.) | 117-119 °C (lit.) |
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service