Key Documents
Z327786
BRAND® graduated pipette, PP, high clarity
capacity 0-2 mL , 0.020 accuracy
Synonym(s):
graduated pipet
About This Item
UNSPSC Code:
41121500
material
polypropylene
description
blue ring mark
feature
graduations
subdivision 0.1 mL
packaging
pack of 12
manufacturer/tradename
BRAND 27610
capacity
0-2 mL , 0.020 accuracy
suction tube O.D.
≤8 mm
General description
Resistant to breakage. Ideal for applications where glass pipettes cannot be used, e.g. in fluoro and silicate chemistry. Exposure to temperatures up to 60°C will not cause permanent exceeding of error limits. Cleaning temperatures below 60°C is recommended to preserve marks and inscriptions
Other Notes
BR27610 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.