Documents
HS120479
Manual pipette controller
blue
About This Item
UNSPSC Code:
41121500
NACRES:
NB.51
manufacturer/tradename
Heathrow Scientific HS120479
volume
1-100 mL
color
blue
General description
Comfortable to hold and easy to use, the manual inline pipette controller was designed with an ergonomic grip and bulb for simple and precise operation. A T-bar thumb lever controls the aspirating and dispensing speeds from serological pipettes from 1 to 100 mL in size.
Operation is simple: Squeeze the bulb to create a vacuum to draw up fluid. Push the control lever backward (upward) to aspirate liquid through a pipette, or push the lever forward (downward) to dispense the liquid. A press of the central bulb on the unit blows-out any remaining fluid if necessary.
Features and Benefits
  • Inline ergonomic design reduces stress with ease of handling and operation
  • Conveniently accommodates either glass or plastic serological or volumetric pipettes ranging from 1 - 100 mL
  • Includes a replaceable .45μm hydrophobic filter to protect your samples and internal components
  • Stay organized with two additional nose cones (green, white) for color identification
  • Polypropylene nozzle and silicone adapter is autoclavable