Skip to Content
Merck
All Photos(1)

Key Documents

U2626

Sigma-Aldrich

Uridine 3′:5′-cyclic monophosphate sodium salt

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H10N2O8PNa
CAS Number:
Molecular Weight:
328.15
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

~98%

form

solid

storage temp.

−20°C

SMILES string

[Na].OC1C2OP(O)(=O)OCC2OC1N3C=CC(=O)NC3=O

InChI

1S/C9H11N2O8P.Na.H/c12-5-1-2-11(9(14)10-5)8-6(13)7-4(18-8)3-17-20(15,16)19-7;;/h1-2,4,6-8,13H,3H2,(H,15,16)(H,10,12,14);;

InChI key

RASOKOFGAKEGDS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service