Skip to Content
Merck
All Photos(1)

Key Documents

SRP0153

Sigma-Aldrich

SMYD3 (full length) Active human

recombinant, expressed in baculovirus infected insect cells, ≥50% (SDS-PAGE)

Synonym(s):

KMT3E, Lysine N-methyltransferase 3E, SET and MYND domain containing 3, ZMYND1, Zinc finger MYND domain-containing protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€650.00

€650.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
€650.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€650.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥50% (SDS-PAGE)

form

aqueous solution

mol wt

50 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMYD3(64754)

General description

Human SMYD3 (GenBank Accession No. BC031010), full length, with N-terminal DDDDK tag (FLAG), MW = 50 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 100 μg/ml flag peptide, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilham M Alshiraihi et al.
International journal of molecular sciences, 21(24) (2020-12-19)
SMYD3 is a lysine methyltransferase that regulates the expression of over 80 genes and is required for the uncontrolled proliferation of most breast, colorectal, and hepatocellular carcinomas. The elimination of SMYD3 restores normal expression patterns of these genes and halts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service