Skip to Content
Merck
All Photos(5)

Documents

SAB3501243

Sigma-Aldrich

Anti-CD81 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD81(975)

General description

CD81 Antibody: CD81 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family, a group of cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that associates with integrins and appears to promote muscle cell fusion and support myotube maintenance. Along with SCARB1, LDL-R, and CLDN1, CD81 has been reported to be an entry factor for the Hepatitis C virus. Finally, recent evidence indicates that the CD81 gene is localized in a tumor-suppressor gene region and is thus a candidate gene for malignancies.

Immunogen

CD81 antibody was raised against a 20 amino acid synthetic peptide near the amino terminus of human CD81.The immunogen is located within amino acids 30 - 80 of CD81.

Application

CD81 antibody can be used for detection of CD81 by Western blot at 2.5 μg/mL. Antibody can also be used for immunohistochemistry starting at 2 μg/mL. For immunofluorescence start at 10 μg/mL.Antibody validated: Western Blot in human samples; Immunohistochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service