Skip to Content
Merck
All Photos(1)

Key Documents

PGR4

Sigma-Aldrich

Isopentenyl Adenosine Immunoassay Detection Kit

Quantitate the levels of Isopentenyl Adenosine in plant tissue

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

Pricing and availability is not currently available.

quality

Quantitate the levels of Isopentenyl Adenosine in plant tissue

application(s)

agriculture

storage temp.

2-8°C

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service