Skip to Content
Merck
All Photos(1)

Key Documents

P6926

Sigma-Aldrich

Phenylhydrazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNH2 · HCl
CAS Number:
Molecular Weight:
144.60
Beilstein:
3594958
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

250-254 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

Cl[H].NNc1ccccc1

InChI

1S/C6H8N2.ClH/c7-8-6-4-2-1-3-5-6;/h1-5,8H,7H2;1H

InChI key

JOVOSQBPPZZESK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Frazer et al.
British journal of haematology, 157(5), 615-626 (2012-03-28)
The BMP/SMAD signalling pathway plays an important role in iron homeostasis, regulating hepcidin expression in response to body iron levels. However, the role of this pathway in the reduction in hepcidin associated with increased erythropoiesis (and secondary iron loading) is
Weiwei Huang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(3), 471-475 (2011-05-03)
A novel colorimetric anion-chemosensor based on 1,10-phenanthroline-2,9-dicarbonyl-p-nitro-phenylhydrazine has been synthesized. Among the different anions tested, it shows the best selectivity towards AcO(-). The addition of acetate causes the color to change from yellow to red, which could be detected with
Jeanne E Hendrickson et al.
Transfusion, 51(11), 2445-2454 (2011-05-17)
Fourteen-day stored red blood cells (RBCs) containing an RBC-specific transgenic antigen (HOD) induce a recipient proinflammatory cytokine storm and are significantly more immunogenic compared to fresh RBCs. Given that recipient mice clear transfused stored RBCs more rapidly than fresh RBCs
Claudia Köthe et al.
Inorganic chemistry, 51(18), 9740-9747 (2012-09-07)
The reaction of [L(tBu)Ni(OEt(2))] (where L(tBu) = [HC(C(t)BuNC(6)H(3)((i)Pr)(2))(2)](-)) with phenylhydrazine leads to the phenylhydrazido(1-) complex [L(tBu)Ni(η(2)-NPhNH(2))] (1) with concomitant formation of H(2). Treatment of 1 with potassium graphite in the presence of crown ether again leads to H(2) evolution and
Dagny L Ulrich et al.
The Journal of biological chemistry, 287(16), 12679-12690 (2012-02-02)
Abcb6 is a mammalian mitochondrial ATP-binding cassette (ABC) transporter that regulates de novo porphyrin synthesis. In previous studies, haploinsufficient (Abcb6(+/-)) embryonic stem cells showed impaired porphyrin synthesis. Unexpectedly, Abcb6(-/-) mice derived from these stem cells appeared phenotypically normal. We hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service