Skip to Content
Merck
All Photos(1)

Key Documents

AV47470

Sigma-Aldrich

Anti-SerPINE1 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PAI, Anti-PAI-1, Anti-PAI1, Anti-PLANH1, Anti-Serpin peptidaSe inhibitor, clade E , member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€454.00

€454.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€454.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€454.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

sheep, human, horse, bovine, rabbit, dog, pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINE1(5054)

General description

SerPINE1 is a serine proteinase inhibitor that blocks fibrinolys by inhibiting tissue plasminogen activator (tPA) and urokinase (uPA). Genetic variations in SERPINE1 have been linked to pre-eclampsia (PE). Studies have reported that targeting SERPINE1 (PAI-1) expression in Alzheimer′s disease may have therapeutic implications. Moreover, MiR-34c regulates SERPINE1 expression in emphysema.
Rabbit Anti-SerPINE1 antibody recognizes human, mouse, rat, zebrafish, bovine, pig, chicken, and canine SERPINE1.

Immunogen

Synthetic peptide directed towards the C terminal region of human SERPINE1

Application

Rabbit Anti-SerPINE1 antibody is suitable for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

SERPINE1 acts as ′bait′ for tissue plasminogen activator, urokinase, and protein C. Its rapid interaction with TPA may function as a major control point in the regulation of fibrinolysis.

Sequence

Synthetic peptide located within the following region: VNESGTVASSSTAVIVSARMAPEEIIMDRPFLFVVRHNPTGTVLFMGQVM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacie M Kutz et al.
Molecular Medicine & Therapeutics, 1(2), 106-106 (2013-07-13)
Accumulation of neurotoxic amyloid peptides (Aβ) in the brain, generated by β-site proteolytic processing of the amyloid precursor protein (APP), is the hallmark pathophysiologic feature of Alzheimer's disease. The plasmin-activating cascade, in which urokinase (uPA) and tissue-type (tPA) plasminogen activators
Santiyagu M Savarimuthu Francis et al.
BMC genomics, 15, 88-88 (2014-02-01)
MicroRNAs (MiRNA) are small non-coding RNAs that regulate gene expression. The aim of this study was to identify miRNAs differentially expressed between mild and moderately emphysematous lung, as well as their functional target mRNAs. Resected lung from patients with COPD
Linlu Zhao et al.
Molecular human reproduction, 19(3), 136-143 (2012-11-28)
The SERPINE1 -675 4G/5G promoter region insertion/deletion polymorphism (rs1799889) has been implicated in the pathogenesis of pre-eclampsia (PE), but the genetic association has been inconsistently replicated. To derive a more precise estimate of the association, a systematic review and meta-analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service