Skip to Content
Merck
All Photos(1)

Key Documents

V0500000

Vindesine sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Vindesine sulfate salt, Desacetylvinblastine amide sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C43H55N5O7 · H2SO4
CAS Number:
Molecular Weight:
852.00
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vindesine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

InChI

1S/C43H55N5O7.H2O4S/c1-6-39(52)21-25-22-42(38(51)55-5,33-27(13-17-47(23-25)24-39)26-11-8-9-12-30(26)45-33)29-19-28-31(20-32(29)54-4)46(3)35-41(28)15-18-48-16-10-14-40(7-2,34(41)48)36(49)43(35,53)37(44)50;1-5(2,3)4/h8-12,14,19-20,25,34-36,45,49,52-53H,6-7,13,15-18,21-24H2,1-5H3,(H2,44,50);(H2,1,2,3,4)/t25-,34+,35-,36-,39+,40-,41-,42+,43+;/m1./s1

InChI key

COFJBSXICYYSKG-FJFFLIEUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vindesine sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernardo A Mainou et al.
mBio, 4(4) (2013-07-04)
Mammalian reovirus binds to cell-surface glycans and junctional adhesion molecule A and enters cells by receptor-mediated endocytosis in a process dependent on β1 integrin. Within the endocytic compartment, reovirus undergoes stepwise disassembly, allowing release of the transcriptionally active viral core
Jianyou Gu et al.
BMC complementary medicine and therapies, 20(1), 214-214 (2020-07-10)
C-Myc overexpression is associated with poor prognosis and aggressive progression of natural killer/T-cell lymphoma (NKTCL). Matrine, a main alkaloid of the traditional Chinese herb Sophora flavescens Ait, has been shown to inhibit cellular proliferation and induce apoptosis of various cancer
R J Cersosimo et al.
Pharmacotherapy, 3(5), 259-274 (1983-09-01)
Vindesine is a new vinca alkaloid antineoplastic agent derived from vinblastine. However, its antineoplastic spectrum more closely resembles that of vincristine. Clinical studies indicate activity against acute leukemia, lung cancer, carcinoma of the breast, squamous cell carcinoma of the esophagus
Manuel Colombo et al.
Annals of work exposures and health, 61(8), 1003-1014 (2017-10-14)
This paper describes a novel wipe sampling and high-performance liquid chromatography/tandem mass spectrometry (HPLC-MS/MS) method capable of simultaneously detecting 10 antineoplastic drugs (5-fluorouracil, oxaliplatin, methotrexate, vindesine, ifosfamide, cyclophosphamide, vincristine, vinblastine, docetaxel, and paclitaxel). The good overall recoveries and sensitivity values
Thierry Jo Molina et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(35), 3996-4003 (2014-11-12)
To determine whether any tumor biomarkers could account for the survival advantage observed in the LNH 03-2B trial among patients with diffuse large B-cell lymphoma (DLBCL) and low-intermediate risk according to the International Prognostic Index when treated with dose-intensive rituximab

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service