Skip to Content
Merck
All Photos(1)

Key Documents

NIST2266

Hopanes and steranes in 2,2,4 trimethylpentane

NIST® SRM® 2266

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 1.2 ML
€1,220.00

€1,220.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 1.2 ML
€1,220.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,220.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

format

multi-component solution

General description

One unit of SRM 2266 contains five ampoules of 2 mL, with each consisting of about 1.2 mL ofsolution.

SRM 2266_cert

SRM 2266_SDS

Application

The SRM is intended for chromatographic instrumentation calibration, used in the determination of hopanes and steranes.

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values of three hopanes and four steranes which is obtained through two methods:
  1. Gravimetric preparation of the solution.
  2. The analytical results determined using gas chromatography.
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Certified for the analytes listed below.
17α(H)-22,29,30-trisnorhopane, 17α(H),21α(H)-30-norhopane, 17α(H),21α(H)-hopane, ααα 20R-cholestane,
ααα 20R-cholestane, ααα 20R 24S-methylcholestane, ααα 20R 24R-ethylcholestane

Reference/Informational value is provided for the following analytes.
17α(H),21α(H)-22R-homohopane, 17α(H),21α(H)- 22S-homohopane, ααα 20R 24R-ethylcholestane

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service