Skip to Content
Merck
All Photos(1)

Documents

NIST2261

Chlorinated pesticides in hexane

NIST® SRM® 2261

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

application(s)

agriculture

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a solution of 15 chlorinated pesticides in hexane and is intended primarily for use in the calibration of gas chromatographic instrumentation used for the determination of chlorinated pesticides. A unit of SRM 2261 consists of five 2 mL ampoules, each containing approximately 1.2 mL of solution. For more information, please refer to the SDS and COA.

SRM 2261_cert

SRM 2261_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Hexachlorobenzene, gamma-HCH, Heptachlor, Heptachlor epoxide, cis-Chlordane, trans-Nonachlor, Dieldrin, Mirex, 2,4-DDE, 4,4-DDE, 2,4-DDD, 2,4-DDT, 4,4-DDT

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service