Skip to Content
Merck
All Photos(1)

Key Documents

MABX8292

Sigma-Aldrich

Anti-Hepatitis B Core Antigen (HBcAg) Antibody, (1mg)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€667.00

€667.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€667.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

€667.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

affinity purified immunoglobulin

clone

2
monoclonal

purified by

affinity chromatography

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG2b

General description

Recombinant full-length HBcAg

Specificity

Core antigen of Hepatitis B virus

Quality

Detect HBcAg using mouse monoclonal Anti-HBcAg, Cat No MABX8292-1KC, validated for use in ELISA


Physical form

Purified mouse monoclonal antibody in buffer containing 10 mM Tris-HCl, pH 8.0, 0.05% NaN3

Storage and Stability

2-8C. For long-term storage, freeze at -20C.

Other Notes

Core Ag of Hepatitis B virus

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service