Skip to Content
Merck
All Photos(1)

Documents

MABX8301

Sigma-Aldrich

Anti-Neisseria gonorrhoeae (NGH) Antibody, (1mg)

clone 1, Chemicon®, from mouse

Synonym(s):

Neisseria gonorrhoeae (NGH)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

1, monoclonal

species reactivity

Neisseria gonorrhoeae, bacteria

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

Neisseria gonorrhoeae is a species of Gram-negative diplococci bacteria that causes a sexually-transmitted genitourinary infection, gonorrhea, in genitals, throat and eyes. Asymptomatic infection is common in males and females but untreated infection may spread to other parts of the body and cause serious disseminated diseases.

Specificity

Neisseria gonorrhoeae. Does not cross-react with: N. meningitidis, N. cinerea, N. lactamica, M. sicca, B. catarrhalis, E. coli, P. mirabilis, Gardnerella vaginalis, or Chlamydia trachomatis

Immunogen

Neisseria gonorrhoeae

Application

Detect Neisseria gonorrhoeae using this mouse monoclonal Anti-NGH, Cat No MABX8301-1KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Infectious Diseases
Suggested antibody pairs MABX8301-1KC (labeling) and MABX8302-1KC (coating)

Quality

Detect Neisseria gonorrhoeae using this mouse monoclonal Anti-NGH, Cat No MABX8301-1KC, validated for use in ELISA

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal antibody in buffer containing 50 mM NaCL, 10 mM PBS, pH 8.0, 0.1% sodium azide (NaN3)

Storage and Stability

2-8C. For long-term storage, freeze at -20C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service