Skip to Content
Merck
All Photos(1)

Documents

MABX7501

Sigma-Aldrich

Anti-human brain natriuretic peptide, clone 51

clone 51, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

51, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

Gene Information

human ... NPPB(4879)

General description

Antibody reacts with Brain Neurotrophic Factor (BNF). BNF is a protein that is released into the blood due to excessive stretching of the heart muscle cells. This is a diagnostic markers that is exclusively used to rule out a heart attack.

Application

Confirmed matched pairing. Capture Antibody MABX7501-1KC; Detector Antibody MABX7630-1KC or MABX7262-1KC

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service