Skip to Content
Merck
All Photos(1)

Key Documents

LSK2ABG20

Millipore

Montage Antibody Purification Kit with PROSEP-G media

Antibody Purification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
Pricing and availability is not currently available.

material

glass bed support (porous)

sterility

non-sterile

manufacturer/tradename

Montage®

parameter

20 mL sample volume (maximum in swinging bucket rotor)

technique(s)

protein purification: suitable (antibody)

bed volume

1.6 mL

particle size

75–125 μm

pH

2 – 9

capacity

>10 mg binding capacity (rabbit IgG)

application(s)

sample preparation

shipped in

ambient

Components

The Montage Antibody Purification Kits include 2 media plugs with PROSEP-G media, 2 spin columns, 4 centrifuge tubes, 4 Steriflip devices, 4 Amicon Ultra devices and optimized buffers to perform up to 20 purifications.

Physical form

Color Code: Yellow

Legal Information

MONTAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Li et al.
mBio, 10(3) (2019-06-06)
Secondary bacterial lung infection by Streptococcus pneumoniae (S. pneumoniae) poses a serious health concern, especially in developing countries. We posit that the emergence of multiantibiotic-resistant strains will jeopardize current treatments in these regions. Deaths arising from secondary infections are more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service