ABS796
Anti-phospho-HER4 Antibody (Tyr1162)
from rabbit, purified by affinity chromatography
Synonym(s):
ERBB4, HER4, Receptor tyrosine-protein kinase erbB-4, Proto-oncogene-like protein c-ErbB-4, Tyrosine kinase-type cell surface receptor HER4, p180erbB4, ERBB4 intracellular domain, s80HER4
Select a Size
Select a Size
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
phosphorylation (pTyr1162)
Gene Information
human ... ERBB4(2066)
General description
Immunogen
Application
Immunocytochemistry Analysis (ICC): A 1:10-50 dilution from a representative lot detected HER4 in MCF-7 cells.
Quality
Western Blotting Analysis (WB): A 1:1000 dilution of this antibody detected HER4 in untreated and EGF treated A431 cell lysates.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service