Skip to Content
Merck
All Photos(1)

Documents

17-317

Sigma-Aldrich

Protein Phosphatase Inhibitor Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
eCl@ss:
32160405
NACRES:
NA.77

availability

available only in USA

Quality Level

suitability

suitable for phosphatase test

General description

The cocktails in this kit are intended for the inhibition of cellular Serine/Threonine protein phosphatases, Tyrosine phosphastases, acid and alkaline phosphatases.

Packaging

Kit capacity: Varies with application

Components

Ser/Thr Protein Phosphatase Inhibitor Cocktail Tyrosine Protein Phosphatase Inhibitor Cocktail

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service