Skip to Content
Merck
All Photos(1)

Documents

8.20825

Sigma-Aldrich

Methyl phenyl sulfide

for synthesis

Synonym(s):

Methyl phenyl sulfide, Thioanisole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8S
CAS Number:
Molecular Weight:
124.20
MDL number:
UNSPSC Code:
12352113
EC Index Number:
202-878-2
NACRES:
NA.22

vapor pressure

38.5 Pa ( 20 °C)

Quality Level

form

liquid

potency

891 mg/kg LD50, oral (Rat)

bp

188 °C/1013 hPa

mp

-17 °C

transition temp

flash point 73 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H8S/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

HNKJADCVZUBCPG-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F

Flash Point(C)

73 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service