Skip to Content
Merck
All Photos(1)

Documents

W502707

Sigma-Aldrich

Succinic acid

FCC

Synonym(s):

Butanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
FEMA Number:
4719
Beilstein:
1754069
EC Number:
Council of Europe no.:
24
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.024

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 184.1091

Assay

99.0-100.5% (by NaOH, titration)

form

crystals

bp

235 °C (lit.)

mp

184-186 °C (lit.)

solubility

water: soluble 83 g/L at 25 °C

density

1.564 g/cm3 at 15 °C

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
Pb: ≤2 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

acidic; sour

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Tannahill et al.
Nature, 496(7444), 238-242 (2013-03-29)
Macrophages activated by the Gram-negative bacterial product lipopolysaccharide switch their core metabolism from oxidative phosphorylation to glycolysis. Here we show that inhibition of glycolysis with 2-deoxyglucose suppresses lipopolysaccharide-induced interleukin-1β but not tumour-necrosis factor-α in mouse macrophages. A comprehensive metabolic map
James H Marden et al.
Evolution; international journal of organic evolution, 67(4), 1105-1115 (2013-04-05)
Oxygen conductance to the tissues determines aerobic metabolic performance in most eukaryotes but has cost/benefit tradeoffs. Here we examine in lowland populations of a butterfly a genetic polymorphism affecting oxygen conductance via the hypoxia-inducible factor (HIF) pathway, which senses intracellular
Susan Jahn et al.
Biochimica et biophysica acta, 1833(12), 2879-2889 (2013-07-28)
To investigate a possible role of the nitrogen-PTS (PTS(Ntr)) in controlling carbon metabolism, we determined the growth of Escherichia coli LJ110 and of isogenic derivatives, mutated in components of the PTS(Ntr), on different carbon sources. The PTS(Ntr) is a set
José Manuel Otero et al.
PloS one, 8(1), e54144-e54144 (2013-01-26)
Saccharomyces cerevisiae is the most well characterized eukaryote, the preferred microbial cell factory for the largest industrial biotechnology product (bioethanol), and a robust commerically compatible scaffold to be exploitted for diverse chemical production. Succinic acid is a highly sought after
Yun Chen et al.
Current opinion in biotechnology, 24(6), 965-972 (2013-04-02)
Bio-based production of chemical building blocks from renewable resources is an attractive alternative to petroleum-based platform chemicals. Metabolic pathway and strain engineering is the key element in constructing robust microbial chemical factories within the constraints of cost effective production. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service