SLW3460/02M
Pyrex® Centrifuge tubes
graduated, conical base, 15 mL
Synonym(s):
Pyrex, SciLabware 3460/02M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
conical base
feature
graduated
manufacturer/tradename
3460/02M
SciLabware
neck I.D.
17 mm
capacity
15 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Centrifuge tube with white enamel graduations
- Graduated 0 - 3ml in 0.1ml divisions and 3 - 10ml in 0.5ml divisions
- Manufactured from Pyrex borosilicate glass
- Can be centrifuged up to 3000 RCF values
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service